This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ALDH1A3 SIRNA Assays ›
Gene Symbol:
Aldh1a3
Gene Name:
aldehyde dehydrogenase 1 family, member A3
Gene Aliases:
Aldh6
Chromosome Location:
Chr.1: 127302920 - 127337828 on Build Rnor_6.0
UniGene ID:
Rn.8297
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 58654

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Aldh1a3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153300.1 NP_695212.1 7, 8 788
GenBank mRNA AB369261.1 - 7, 8 788
AF434845.1 - 7, 8 788
BC166415.1 - 845 9025597

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

kidney development
optic cup morphogenesis involved in camera-type eye development
retinoic acid biosynthetic process
locomotory behavior
nucleus accumbens development
pituitary gland development
embryonic camera-type eye development
inner ear morphogenesis
response to drug
retinol metabolic process
retinoic acid metabolic process
retinal metabolic process
positive regulation of apoptotic process
embryonic eye morphogenesis
neuromuscular process controlling balance
oxidation-reduction process
righting reflex
olfactory pit development
face development
retinal dehydrogenase activity
aldehyde dehydrogenase (NAD) activity
aldehyde dehydrogenase [NAD(P)+] activity
protein homodimerization activity
thyroid hormone binding
NAD+ binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0