This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FABP5 SIRNA Assays ›
Gene Symbol:
Fabp5
Gene Name:
fatty acid binding protein 5, epidermal
Gene Aliases:
C-FABP, DA11, E-FABP
Chromosome Location:
Chr.2: 93981659 - 93985358 on Build Rnor_6.0
UniGene ID:
Rn.98269
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 58433

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fabp5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145878.1 NP_665885.1 3 353
XM_006232136.2 XP_006232198.1 2 212
GenBank mRNA DQ255913.1 - 2 211
FQ210725.1 - 3 363
FQ228381.1 - 3 365
S69874.1 - 3 355
S83247.1 - 3 370
U13253.1 - 3 353

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

glucose metabolic process
lipid metabolic process
phosphatidylcholine biosynthetic process
transport
response to wounding
glucose transport
transporter activity
fatty acid binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0