This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GPR37L1 SIRNA Assays ›
Gene Symbol:
Gpr37l1
Gene Name:
G protein-coupled receptor 37-like 1
Gene Aliases:
-
Chromosome Location:
Chr.13: 51985698 - 51992547 on Build Rnor_6.0
UniGene ID:
Rn.144679
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 58340

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gpr37l1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145784.2 NP_665727.2 1 216
GenBank mRNA AF087947.1 - 282
BC168215.1 - 1 216 7313732

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
positive regulation of cerebellar granule cell precursor proliferation
positive regulation of MAPK cascade
negative regulation of neuron differentiation
negative regulation of smoothened signaling pathway
negative regulation of astrocyte differentiation
negative regulation of hydrogen peroxide-induced cell death
G-protein coupled receptor activity
G-protein coupled peptide receptor activity
peptide binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0