This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ACOX2 SIRNA Assays ›
Gene Symbol:
Acox2
Gene Name:
acyl-CoA oxidase 2, branched chain
Gene Aliases:
THCCox
Chromosome Location:
Chr.15: 18449304 - 18481472 on Build Rnor_6.0
UniGene ID:
Rn.10622
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 58328

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Acox2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145770.2 NP_665713.2 3 440
XM_006251743.2 XP_006251805.1 4 525
XM_006251744.2 XP_006251806.1 3 396
XM_006251745.2 XP_006251807.1 4 443
GenBank mRNA X95189.1 - 3 439

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

fatty acid metabolic process
fatty acid beta-oxidation
acyl-CoA dehydrogenase activity
acyl-CoA oxidase activity
receptor binding
fatty acid binding
3alpha,7alpha,12alpha-trihydroxy-5beta-cholestanoyl-CoA 24-hydroxylase activity
flavin adenine dinucleotide binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0