This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RNH1 SIRNA Assays ›
Gene Symbol:
Rnh1
Gene Name:
ribonuclease/angiogenin inhibitor 1
Gene Aliases:
Rnh
Chromosome Location:
Chr.1: 214151337 - 214163807 on Build Rnor_6.0
UniGene ID:
Rn.3756
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 58134

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rnh1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270762.1 NP_001257691.1 6 762
NM_001270763.1 NP_001257692.1 6 768
NM_139105.3 NP_620805.2 6 777
XM_008759970.1 XP_008758192.1 6 1326
GenBank mRNA AB296102.1 - 5 644
BC070501.1 - 6 684 7096144
FQ233708.1 - 6 702
FQ234889.1 - 6 701
X62528.1 - 6 762

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of catalytic activity
regulation of angiogenesis
ribonuclease inhibitor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header