This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CRISPLD2 SIRNA Assays ›
Gene Symbol:
Crispld2
Gene Name:
cysteine-rich secretory protein LCCL domain containing 2
Gene Aliases:
Lcrisp2, Lgl1
Chromosome Location:
Chr.19: 52647035 - 52705118 on Build Rnor_6.0
UniGene ID:
Rn.4346
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 57635

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Crispld2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138518.2 NP_612527.2 4, 5 686
XM_006255708.2 XP_006255770.1 5, 6 809
GenBank mRNA AF109674.1 - 4, 5 701
CB732240.1 - 2, 3 278

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

extracellular matrix organization
lung development
face morphogenesis
glycosaminoglycan binding
heparin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header