This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SPATA19 SIRNA Assays ›
Gene Symbol:
Spata19
Gene Name:
spermatogenesis associated 19
Gene Aliases:
Spas1, spergen-1
Chromosome Location:
Chr.8: 28454938 - 28460645 on Build Rnor_6.0
UniGene ID:
Rn.51863
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 57394

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Spata19
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134384.1 NP_599211.1 1, 2 100
GenBank mRNA AB057408.1 - 1, 2 100
BC081729.1 - 1, 2 117 7132055

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_029299, AB120716, AK005610.

Gene Ontology Categories:

Function(s)

multicellular organism development
spermatogenesis
cell differentiation

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0