This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NDEL1 SIRNA Assays ›
Gene Symbol:
Ndel1
Gene Name:
nudE neurodevelopment protein 1-like 1
Gene Aliases:
NUDE2, Nudel
Chromosome Location:
Chr.10: 55411888 - 55456654 on Build Rnor_6.0
UniGene ID:
Rn.2947
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 56957

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ndel1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133320.1 NP_579854.1 3 222
XM_006246571.2 XP_006246633.1 3 631
XM_006246572.2 XP_006246634.1 3 624
XM_006246573.2 XP_006246635.1 3 372
XM_006246574.2 XP_006246636.1 3 257
XM_006246575.2 XP_006246637.1 3 735
XM_006246576.2 XP_006246638.1 3 625
XM_006246578.2 XP_006246640.1 3 371
GenBank mRNA AY008298.1 - 3 222
BC070909.1 - 3 312 7103650

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_023668, AF323918, AK146029, AF290472, BC046796, and others.

Gene Ontology Categories:

Function(s) Process(es)

establishment of mitotic spindle orientation
microtubule cytoskeleton organization
neuron migration
inner cell mass cell proliferation
proteolysis
microtubule nucleation
chromosome segregation
mitotic centrosome separation
retrograde axonal transport
regulation of neuron projection development
cell migration
cerebral cortex radially oriented cell migration
central nervous system neuron axonogenesis
neuron projection development
positive regulation of axon extension
vesicle transport along microtubule
positive regulation of axon regeneration
nuclear envelope disassembly
establishment of chromosome localization
centrosome localization
neurofilament cytoskeleton organization
canonical Wnt signaling pathway
neuron projection extension
protein binding
microtubule binding
protein complex binding
alpha-tubulin binding
beta-tubulin binding
oligopeptidase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0