This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MAP2K2 SIRNA Assays ›
Gene Symbol:
Map2k2
Gene Name:
mitogen activated protein kinase kinase 2
Gene Aliases:
Prkmk2
Chromosome Location:
Chr.7: 11458971 - 11478520 on Build Rnor_6.0
UniGene ID:
Rn.82693
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 56927

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Map2k2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133283.1 NP_579817.1 2 241
XM_006240987.2 XP_006241049.1 2 428
GenBank mRNA BC126084.1 - 2 339 8376547
D14592.1 - 2 212
L14936.1 - 2 241

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
activation of MAPK activity
protein phosphorylation
peptidyl-tyrosine phosphorylation
regulation of stress-activated MAPK cascade
peptidyl-serine autophosphorylation
regulation of axon regeneration
positive regulation of axonogenesis
lung morphogenesis
trachea formation
epithelial cell proliferation involved in lung morphogenesis
ERK1 and ERK2 cascade
positive regulation of protein serine/threonine kinase activity
regulation of Golgi inheritance
positive regulation of cell motility
regulation of early endosome to late endosome transport
protein serine/threonine kinase activity
MAP kinase kinase activity
protein serine/threonine/tyrosine kinase activity
protein tyrosine kinase activity
protein binding
ATP binding
PDZ domain binding
protein complex scaffold
protein serine/threonine kinase activator activity
scaffold protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0