This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ADCYAP1R1 SIRNA Assays ›
Gene Symbol:
Adcyap1r1
Gene Name:
adenylate cyclase activating polypeptide 1 receptor 1
Gene Aliases:
PAC1-R, PACAP-R1, PACAP-R1A, PACAPR1, PACAPR1A
Chromosome Location:
Chr.4: 85662892 - 85711696 on Build Rnor_6.0
UniGene ID:
Rn.234543
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 56912

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Adcyap1r1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270579.1 NP_001257508.1 4 579
NM_001270580.1 NP_001257509.1 4 579
NM_001270581.1 NP_001257510.1 5 691
NM_001270582.1 NP_001257511.1 4 579
NM_001270583.1 NP_001257512.1 4 579
NM_133511.2 NP_598195.1 4 579
XM_008762870.1 XP_008761092.1 3 325
GenBank mRNA D14908.1 - 5 571
D14909.1 - 4 369
D16465.1 - 4 579
L16680.1 - 3 272
Z23272.1 - 3 332
Z23273.1 - 3 332
Z23274.1 - 3 332
Z23275.1 - 3 332
Z23279.1 - 3 332
Z23282.1 - 4 635

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001025372, NM_007407, AK042820, AK081121, AK147291, and others.

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
activation of phospholipase C activity
spermatogenesis
positive regulation of calcium ion transport into cytosol
cAMP-mediated signaling
cell differentiation
positive regulation of cAMP biosynthetic process
response to estradiol
multicellular organismal response to stress
response to drug
response to ethanol
development of primary female sexual characteristics
negative regulation of cell death
positive regulation of inositol phosphate biosynthetic process
vasoactive intestinal polypeptide receptor activity
adenylate cyclase binding
ADP-ribosylation factor binding
neuropeptide binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0