This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other A1CF SIRNA Assays ›
Gene Symbol:
A1cf
Gene Name:
APOBEC1 complementation factor
Gene Aliases:
A1cft, Acf, Apobec-1
Chromosome Location:
Chr.1: 250426021 - 250514245 on Build Rnor_6.0
UniGene ID:
Rn.162520
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 56884

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: A1cf
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133400.1 NP_596891.1 2 192
XM_006231272.2 XP_006231334.1 2 363
GenBank mRNA AF290984.1 - 2 192
AF442133.1 - 2 192
AF442134.1 - 2 192
AY028945.1 - 1 90

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_001198820, NM_001198818, NM_138932, NM_014576, NM_001198819, and others.
This siRNA also targets the following transcripts in Mouse: NM_001081074, AY566862, AY566863, AY566864, BC089622.

Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
mRNA localization resulting in posttranscriptional regulation of gene expression
cytidine to uridine editing
mRNA modification
protein stabilization
nucleotide binding
nucleic acid binding
double-stranded RNA binding
single-stranded RNA binding
mRNA binding
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0