This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PDE7B SIRNA Assays ›
Gene Symbol:
Pde7b
Gene Name:
phosphodiesterase 7B
Gene Aliases:
-
Chromosome Location:
Chr.1: 15889845 - 16203909 on Build Rnor_6.0
UniGene ID:
Rn.234527
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 56656

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pde7b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080894.2 NP_543170.1 4 312
XM_008758591.1 XP_008756813.1 3 632
XM_008758592.1 XP_008756814.1 3 216
GenBank mRNA AB057409.1 - 4 312
AB057410.1 - 3 180
AB057411.1 - 5 281

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

metabolic process
cAMP-mediated signaling
3',5'-cyclic-AMP phosphodiesterase activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0