This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LEF1 SIRNA Assays ›
Gene Symbol:
Lef1
Gene Name:
lymphoid enhancer binding factor 1
Gene Aliases:
-
Chromosome Location:
Chr.2: 236232117 - 236345061 on Build Rnor_6.0
UniGene ID:
Rn.228611
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 56602

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Lef1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130429.1 NP_569113.1 1 104
XM_006233302.2 XP_006233364.1 1 1423
XM_006233303.2 XP_006233365.1 1 1422
XM_006233305.2 XP_006233367.1 1 1423
XM_006233306.2 XP_006233368.1 1 1422
XM_008761489.1 XP_008759711.1 1 1425
XM_008761490.1 XP_008759712.1 1 1425
GenBank mRNA AF198533.1 - 1 104

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
patterning of blood vessels
osteoblast differentiation
neural crest cell migration
somitogenesis
kidney development
epithelial to mesenchymal transition
vasculature development
sprouting angiogenesis
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
cell adhesion
positive regulation of cell proliferation
response to lithium ion
positive regulation of gene expression
positive regulation of epithelial to mesenchymal transition
response to organic cyclic compound
Wnt signaling pathway
regulation of striated muscle tissue development
dentate gyrus development
hippocampus development
hypothalamus development
forebrain radial glial cell differentiation
forebrain neuroblast division
forebrain neuron differentiation
formation of radial glial scaffolds
regulation of cell-cell adhesion
negative regulation of cell-cell adhesion
positive regulation of cell-cell adhesion
regulation of Wnt signaling pathway
neutrophil differentiation
positive regulation of cell growth
embryonic limb morphogenesis
positive regulation of cell migration
BMP signaling pathway
positive regulation of granulocyte differentiation
mammary gland development
organ regeneration
negative regulation of interleukin-13 production
negative regulation of interleukin-4 production
negative regulation of interleukin-5 production
T cell receptor V(D)J recombination
B cell proliferation
odontogenesis of dentin-containing tooth
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of DNA binding
steroid hormone mediated signaling pathway
tongue development
skin development
positive regulation by host of viral transcription
histone H3 acetylation
histone H4 acetylation
T-helper 1 cell differentiation
negative regulation of striated muscle tissue development
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
alpha-beta T cell differentiation
eye pigmentation
paraxial mesoderm formation
cell development
muscle fiber development
sensory perception of taste
palate development
anatomical structure regression
canonical Wnt signaling pathway
face morphogenesis
cell chemotaxis
apoptotic process involved in morphogenesis
chorio-allantoic fusion
trachea gland development
cellular response to cytokine stimulus
cellular response to interleukin-4
positive regulation of cell proliferation in bone marrow
negative regulation of apoptotic process in bone marrow
odontoblast differentiation
negative regulation of estrogen receptor binding
positive regulation of cell cycle process
negative regulation of canonical Wnt signaling pathway
apoptotic process involved in patterning of blood vessels
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
protein binding
beta-catenin binding
transcription factor binding
DNA binding, bending
estrogen receptor activity
estrogen receptor binding
enhancer binding
histone binding
cysteine-type endopeptidase inhibitor activity involved in apoptotic process
sequence-specific DNA binding
transcription regulatory region DNA binding
gamma-catenin binding
armadillo repeat domain binding
C2H2 zinc finger domain binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0