This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MSMO1 SIRNA Assays ›
Gene Symbol:
Msmo1
Gene Name:
methylsterol monooxygenase 1
Gene Aliases:
Sc4mol
Chromosome Location:
Chr.16: 26859441 - 26875880 on Build Rnor_6.0
UniGene ID:
Rn.7167
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 56553

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Msmo1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080886.1 NP_543162.1 3 371
GenBank mRNA BC063155.1 - 3 358 6918081
D50559.1 - 3 371

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_025436, AK169017, BC006802, AK045059, AK005090, and others.

Gene Ontology Categories:

Function(s) Process(es)

fatty acid biosynthetic process
sterol biosynthetic process
oxidation-reduction process
C-4 methylsterol oxidase activity
iron ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0