This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other AP2B1 SIRNA Assays ›
Gene Symbol:
Ap2b1
Gene Name:
adaptor-related protein complex 2, beta 1 subunit
Gene Aliases:
-
Chromosome Location:
Chr.10: 70516544 - 70621973 on Build Rnor_6.0
UniGene ID:
Rn.56138
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 56528

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ap2b1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080583.1 NP_542150.1 11 1561
XM_008767935.1 XP_008766157.1 12 2318
XM_008767936.1 XP_008766158.1 11 1532
XM_008767937.1 XP_008766159.1 10 1447
XM_008767938.1 XP_008766160.1 10 1464
XM_008767939.1 XP_008766161.1 11 2300
GenBank mRNA BC103481.1 - 12 1530 7929959
M34176.1 - 11 1493
M77246.1 - 11 1561

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
endocytosis
clathrin coat assembly
protein binding
protein transporter activity
clathrin binding
protein complex binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0