This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PHKG2 SIRNA Assays ›
Gene Symbol:
Phkg2
Gene Name:
phosphorylase kinase, gamma 2 (testis)
Gene Aliases:
-
Chromosome Location:
Chr.1: 199019304 - 199032047 on Build Rnor_6.0
UniGene ID:
Rn.11153
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 56439

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Phkg2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080584.4 NP_542151.1 4 687
XM_008759855.1 XP_008758077.1 5 510
XM_008759856.1 XP_008758078.1 5 628
GenBank mRNA BC107906.1 - 4 547
BC128715.1 - 4 616 7456955
M73808.1 - 4 687

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_026888, BC145734, AK005277, BC138913.

Gene Ontology Categories:

Function(s) Process(es)

glycogen metabolic process
glycogen biosynthetic process
protein phosphorylation
phosphorylase kinase activity
calmodulin binding
ATP binding
enzyme binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0