This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DNM1 SIRNA Assays ›
Gene Symbol:
Dnm1
Gene Name:
dynamin 1
Gene Aliases:
Dnm
Chromosome Location:
Chr.3: 11338081 - 11382043 on Build Rnor_6.0
UniGene ID:
Rn.10354
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 56349

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dnm1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080689.4 NP_542420.1 4 552
GenBank mRNA X54531.1 - 4 552

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010065, AK011651, L31397, AK144142, AK220483, and others.

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor internalization
endocytosis
receptor-mediated endocytosis
endosome organization
sensory perception of sound
metabolic process
adult locomotory behavior
receptor internalization
protein tetramerization
synaptic transmission, GABAergic
clathrin-mediated endocytosis
toxin transport
positive regulation of synaptic vesicle recycling
GTPase activity
protein binding
GTP binding
protein C-terminus binding
protein kinase binding
D2 dopamine receptor binding
protein complex binding
identical protein binding
poly(A) RNA binding
protein dimerization activity
nitric-oxide synthase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header