This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MYLK2 SIRNA Assays ›
Gene Symbol:
Mylk2
Gene Name:
myosin light chain kinase 2
Gene Aliases:
Klmc, Mlck
Chromosome Location:
Chr.3: 148386185 - 148397851 on Build Rnor_6.0
UniGene ID:
Rn.81191
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 56312

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mylk2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057209.1 NP_476557.1 2 143
GenBank mRNA J03886.1 - 2 143

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
water transport
muscle contraction
regulation of muscle contraction
striated muscle contraction
neuromuscular synaptic transmission
positive regulation of gene expression
skeletal muscle satellite cell differentiation
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
positive regulation of fast-twitch skeletal muscle fiber contraction
regulation of muscle filament sliding
skeletal muscle cell differentiation
regulation of MAPK cascade
protein autophosphorylation
regulation of neuronal synaptic plasticity
synaptic vesicle transport
positive regulation of cellular component movement
regulation of cell cycle
cardiac muscle tissue morphogenesis
protein serine/threonine kinase activity
myosin light chain kinase activity
calmodulin binding
ATP binding
myosin light chain binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0