This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GPR37 SIRNA Assays ›
Gene Symbol:
Gpr37
Gene Name:
G protein-coupled receptor 37
Gene Aliases:
Ednrbl
Chromosome Location:
Chr.4: 51822163 - 51844228 on Build Rnor_6.0
UniGene ID:
Rn.28035
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 56307

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gpr37
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057201.1 NP_476549.1 1 859
GenBank mRNA AF087946.1 - 1 859
BC087728.1 - 1 925 7320414

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
locomotion involved in locomotory behavior
dopamine biosynthetic process
positive regulation of MAPK cascade
positive regulation of dopamine metabolic process
negative regulation of hydrogen peroxide-induced cell death
G-protein coupled peptide receptor activity
Hsp70 protein binding
heat shock protein binding
ubiquitin protein ligase binding
peptide binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0