This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PFKFB3 SIRNA Assays ›
Gene Symbol:
Pfkfb3
Gene Name:
6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 3
Gene Aliases:
-
Chromosome Location:
Chr.17: 70632801 - 70713736 on Build Rnor_6.0
UniGene ID:
Rn.10791
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 56276

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pfkfb3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057135.1 NP_476476.1 5 814
XM_006254192.2 XP_006254254.1 5 1403
XM_006254193.2 XP_006254255.1 5 1403
XM_006254195.2 XP_006254257.1 5 1403
XM_006254196.2 XP_006254258.1 5 1403
XM_006254197.2 XP_006254259.1 5 1402
XM_006254198.2 XP_006254260.1 5 1402
XM_008771850.1 XP_008770072.1 5 1403
XM_008771851.1 XP_008770073.1 5 430
GenBank mRNA AB006710.1 - 4 229
D87240.1 - 5 814
D87241.1 - 5 814
D87242.1 - 5 814
D87243.1 - 5 814
D87244.1 - 5 814
D87245.1 - 5 814
D87246.1 - 5 814
D87247.1 - 5 814
EU034674.1 - 5 409

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

fructose metabolic process
fructose 2,6-bisphosphate metabolic process
regulation of glycolytic process
brain development
dephosphorylation
carbohydrate phosphorylation
6-phosphofructo-2-kinase activity
fructose-2,6-bisphosphate 2-phosphatase activity
ATP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0