This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other EFNA5 SIRNA Assays ›
Gene Symbol:
Efna5
Gene Name:
ephrin A5
Gene Aliases:
Lerk7
Chromosome Location:
Chr.9: 110053843 - 110329880 on Build Rnor_6.0
UniGene ID:
Rn.10714
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 55993

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Efna5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053903.1 NP_446355.1 2 443
XM_006245593.2 XP_006245655.1 2 636
GenBank mRNA U69279.1 - 2 443

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010109, NM_207654, U90665, BC040218, U90664.

Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
axon guidance
axonal fasciculation
central nervous system development
brain development
regulation of cell-cell adhesion
retinal ganglion cell axon guidance
regulation of actin cytoskeleton organization
ephrin receptor signaling pathway
positive regulation of peptidyl-tyrosine phosphorylation
negative chemotaxis
regulation of focal adhesion assembly
regulation of insulin secretion involved in cellular response to glucose stimulus
regulation of microtubule cytoskeleton organization
chemorepellent activity
ephrin receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0