This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other BCL2L10 SIRNA Assays ›
Gene Symbol:
Bcl2l10
Gene Name:
BCL2-like 10 (apoptosis facilitator)
Gene Aliases:
-
Chromosome Location:
Chr.8: 82288705 - 82294750 on Build Rnor_6.0
UniGene ID:
Rn.67084
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 55694

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Bcl2l10
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053733.1 NP_446185.1 1 342
GenBank mRNA AY029163.1 - 1 342

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
intrinsic apoptotic signaling pathway in response to DNA damage
positive regulation of apoptotic process
negative regulation of apoptotic process
extrinsic apoptotic signaling pathway in absence of ligand
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
negative regulation of intrinsic apoptotic signaling pathway
molecular_function
protein homodimerization activity
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0