This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC7A10 SIRNA Assays ›
Gene Symbol:
Slc7a10
Gene Name:
solute carrier family 7 (neutral amino acid transporter light chain, asc system), member 10
Gene Aliases:
Asc-1
Chromosome Location:
Chr.1: 91432952 - 91449504 on Build Rnor_6.0
UniGene ID:
Rn.128083
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 55689

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc7a10
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053726.2 NP_446178.2 4, 5 713
XM_006228873.2 XP_006228935.1 4, 5 791
XM_006228874.2 XP_006228936.1 4, 5 791
GenBank mRNA AB126813.1 - 4, 5 713
BC127467.1 - 4, 5 684 7313174

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_017394, BC054765, AK031867, AB026688, AK005282.

Gene Ontology Categories:

Function(s) Process(es)

amino acid transmembrane transport
neutral amino acid transport
L-serine transport
D-alanine transport
D-serine transport
neutral amino acid transmembrane transporter activity
L-serine transmembrane transporter activity
D-serine transmembrane transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0