This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NTN1 SIRNA Assays ›
Gene Symbol:
Ntn1
Gene Name:
netrin 1
Gene Aliases:
netrin-1
Chromosome Location:
Chr.10: 54761925 - 54982039 on Build Rnor_6.0
UniGene ID:
Rn.17851
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 55608

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ntn1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053731.2 NP_446183.2 5 1449
XM_006246567.2 XP_006246629.1 6 1801
XM_006246568.2 XP_006246630.1 6 1635
GenBank mRNA AY028417.1 - 5 1449

Back To Top

More Information


Gene Ontology Categories:

Function(s)

neuron migration
apoptotic process
axonogenesis
axon guidance
positive regulation of cell proliferation
single organismal cell-cell adhesion
regulation of cell migration
negative regulation of axon extension
mammary gland development
anterior/posterior axon guidance
establishment of nucleus localization
inner ear morphogenesis
positive regulation of axon extension
mammary gland duct morphogenesis

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0