This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FLT4 SIRNA Assays ›
Gene Symbol:
Flt4
Gene Name:
fms-related tyrosine kinase 4
Gene Aliases:
Vegfr3
Chromosome Location:
Chr.10: 35078782 - 35120296 on Build Rnor_6.0
UniGene ID:
Rn.81043
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 55566

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Flt4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053652.1 NP_446104.1 3 245
GenBank mRNA AF402785.1 - 3 245
AF402786.1 - 3 245

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

positive regulation of protein phosphorylation
positive regulation of endothelial cell proliferation
vasculature development
lymph vessel development
lymphangiogenesis
sprouting angiogenesis
positive regulation of cell proliferation
positive regulation of vascular endothelial growth factor production
positive regulation of endothelial cell migration
peptidyl-tyrosine phosphorylation
positive regulation of cell growth
cellular response to vascular endothelial growth factor stimulus
vascular endothelial growth factor signaling pathway
negative regulation of apoptotic process
positive regulation of MAPK cascade
positive regulation of JNK cascade
protein autophosphorylation
vascular endothelial growth factor receptor signaling pathway
blood vessel morphogenesis
regulation of blood vessel remodeling
positive regulation of ERK1 and ERK2 cascade
positive regulation of protein kinase C signaling
transmembrane receptor protein tyrosine kinase activity
vascular endothelial growth factor-activated receptor activity
ATP binding
growth factor binding
protein phosphatase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0