This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PDLIM3 SIRNA Assays ›
Gene Symbol:
Pdlim3
Gene Name:
PDZ and LIM domain 3
Gene Aliases:
Actn2lp, Alp
Chromosome Location:
Chr.16: 49543140 - 49574357 on Build Rnor_6.0
UniGene ID:
Rn.13361
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 55565

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pdlim3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053650.1 NP_446102.1 2 292
XM_006253119.2 XP_006253181.1 2 327
XM_006253120.2 XP_006253182.1 2 325
GenBank mRNA AF002281.1 - 2 310
BC081703.1 - 2 284 7193873
FQ215471.1 - 2 307

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

actin filament organization
heart development
skeletal muscle tissue development
biological_process
cytoskeletal protein binding
zinc ion binding
structural constituent of muscle
actinin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0