This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SARDH SIRNA Assays ›
Gene Symbol:
Sardh
Gene Name:
sarcosine dehydrogenase
Gene Aliases:
-
Chromosome Location:
Chr.3: 5737202 - 5802146 on Build Rnor_6.0
UniGene ID:
Rn.89832
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 55477

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sardh
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053664.2 NP_446116.1 4 666
XM_006233731.2 XP_006233793.1 5 688
XM_006233732.2 XP_006233794.1 5 1124
XM_006233733.2 XP_006233795.1 5 690
GenBank mRNA AF067650.1 - 4 701
FQ218777.1 - 4 962
L79910.1 - 3 411
U62481.1 - 3 411

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

glycine biosynthetic process
tetrahydrofolate interconversion
choline catabolic process
oxidation-reduction process
folic acid binding
sarcosine dehydrogenase activity
oxidoreductase activity
flavin adenine dinucleotide binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0