This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PSMB1 SIRNA Assays ›
Gene Symbol:
Psmb1
Gene Name:
proteasome (prosome, macropain) subunit, beta type 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 57470878 - 57491359 on Build Rnor_6.0
UniGene ID:
Rn.6016
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 55425

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Psmb1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053590.1 NP_446042.1 5, 6 563
XM_006227981.1 XP_006228043.1 7, 8 818
GenBank mRNA BC058455.1 - 5, 6 575 6922317
FQ211516.1 - 5, 6 577
FQ214442.1 - 5, 6 578
FQ214726.1 - 7, 8 818
FQ218426.1 - 5, 6 548
FQ225328.1 - 5, 6 578
FQ229756.1 - 5, 6 578
FQ230094.1 - 5, 6 578
X52783.1 - 5, 6 563

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011185, AK167647, U60824, X80686, BC018351, and others.

Gene Ontology Categories:

Function(s) Process(es)

proteolysis involved in cellular protein catabolic process
threonine-type endopeptidase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0