This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PSMB7 SIRNA Assays ›
Gene Symbol:
Psmb7
Gene Name:
proteasome (prosome, macropain) subunit, beta type 7
Gene Aliases:
Z
Chromosome Location:
Chr.3: 22890072 - 22951718 on Build Rnor_6.0
UniGene ID:
Rn.3846
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 55199

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Psmb7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053532.1 NP_445984.1 2, 3 171
GenBank mRNA AF285103.1 - 2, 3 171
BC060551.1 - 2, 3 181 6922407
FQ210403.1 - 1, 2 64
FQ212413.1 - 1, 2 64
FQ212825.1 - 1, 2 64
FQ213190.1 - 1, 2 64
FQ213755.1 - 1, 2 64
FQ215265.1 - 1, 2 64
FQ216419.1 - 1, 2 64
FQ218632.1 - 1, 2 64
FQ219250.1 - 1, 2 64
FQ219494.1 - 1, 2 64
FQ219779.1 - 1, 2 64
FQ220023.1 - 1, 2 64
FQ224099.1 - 2, 3 177
FQ229108.1 - 1, 2 64
FQ229165.1 - 1, 2 64
FQ229238.1 - 1, 2 64
FQ229410.1 - 1, 2 64
FQ229570.1 - 1, 2 64
FQ229620.1 - 1, 2 64
FQ229735.1 - 1, 2 64
FQ229750.1 - 1, 2 64
FQ230042.1 - 1, 2 64
FQ232691.1 - 1, 2 64

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

proteolysis involved in cellular protein catabolic process
threonine-type endopeptidase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0