siRNA Details
Gene Symbol: Cnga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053497.1 | NP_445949.1 | 3 | 234 | ||
GenBank mRNA | U93851.1 | - | 3 | 234 |

Gene Symbol: Cnga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053497.1 | NP_445949.1 | 3 | 234 | ||
GenBank mRNA | U93851.1 | - | 3 | 234 |