This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TRPC4 SIRNA Assays ›
Gene Symbol:
Trpc4
Gene Name:
transient receptor potential cation channel, subfamily C, member 4
Gene Aliases:
Trp4, Trrp4
Chromosome Location:
Chr.2: 143433183 - 143605757 on Build Rnor_6.0
UniGene ID:
Rn.10853
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 55049

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Trpc4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083115.1 NP_001076584.1 1 170
NM_080396.1 NP_536321.1 1 170
XM_006232356.2 XP_006232418.1 2 494
XM_008761008.1 XP_008759230.1 2 494
GenBank mRNA AB008889.2 - 2 283
AF288407.1 - 1 186
AF288408.1 - 1 186
AF421363.1 - 1 170
AF421364.1 - 1 170
AF421365.1 - 1 170
AF421366.1 - 1 170
AF421367.1 - 1 170
AF421368.1 - 1 170
FQ228645.1 - 2 389

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

calcium ion transport
gamma-aminobutyric acid secretion
oligodendrocyte differentiation
regulation of calcium ion transport
calcium ion import
calcium ion transmembrane transport
calcium channel activity
beta-catenin binding
store-operated calcium channel activity
cadherin binding
inositol 1,4,5 trisphosphate binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0