This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HERPUD1 SIRNA Assays ›
Gene Symbol:
Herpud1
Gene Name:
homocysteine-inducible, endoplasmic reticulum stress-inducible, ubiquitin-like domain member 1
Gene Aliases:
Sup
Chromosome Location:
Chr.19: 11046903 - 11057570 on Build Rnor_6.0
UniGene ID:
Rn.4028
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 55015

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Herpud1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053523.1 NP_445975.1 2 255
XM_006255133.2 XP_006255195.1 2 571
GenBank mRNA AB033771.1 - 2 255
BC061849.1 - 2 277 5621877

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_001272103, NM_014685, NM_001010989, AK292088, BC000086, and others.
This siRNA also targets the following transcripts in Mouse: NM_022331, BC013523, AK159500, AK081688, AK172874, and others.

Gene Ontology Categories:

Function(s) Process(es)

ubiquitin-dependent protein catabolic process
cellular calcium ion homeostasis
response to unfolded protein
endoplasmic reticulum unfolded protein response
regulation of protein ubiquitination
positive regulation of protein binding
endoplasmic reticulum calcium ion homeostasis
response to endoplasmic reticulum stress
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
positive regulation of ER-associated ubiquitin-dependent protein catabolic process
negative regulation of intrinsic apoptotic signaling pathway
ion channel binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0