This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FGF13 SIRNA Assays ›
Gene Symbol:
Fgf13
Gene Name:
fibroblast growth factor 13
Gene Aliases:
FGF-13
Chromosome Location:
Chr.X: 142053648 - 142571077 on Build Rnor_6.0
UniGene ID:
Rn.15709
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 54950

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fgf13
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053428.1 NP_445880.1 2 226
XM_006257592.1 XP_006257654.1 2 494
XM_008773572.1 XP_008771794.1 3 402
XM_008773573.1 XP_008771795.1 3 464
GenBank mRNA AF271786.1 - 2 226

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010200, AK141848, U66202, BC018238, AF020737.

Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
neuron migration
apoptotic process
negative regulation of microtubule depolymerization
learning
memory
cell proliferation
hippocampus development
cerebral cortex cell migration
establishment of neuroblast polarity
positive regulation of protein kinase activity
microtubule polymerization
negative regulation of collateral sprouting
protein localization to plasma membrane
microtubule binding
growth factor activity
sodium channel regulator activity
protein kinase activator activity
ion channel binding
beta-tubulin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0