This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HOMER3 SIRNA Assays ›
Gene Symbol:
Homer3
Gene Name:
homer homolog 3 (Drosophila)
Gene Aliases:
Vesl-3
Chromosome Location:
Chr.16: 20880447 - 20890966 on Build Rnor_6.0
UniGene ID:
Rn.144573
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 54876

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Homer3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053310.1 NP_445762.1 6 619
XM_006252890.2 XP_006252952.1 7 745
XM_006252891.2 XP_006252953.1 7 742
XM_008771094.1 XP_008769316.1 7 743
XM_008771095.1 XP_008769317.1 7 743
GenBank mRNA AB020879.2 - 6 619

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled glutamate receptor signaling pathway
regulation of store-operated calcium entry
protein C-terminus binding
protein domain specific binding
G-protein coupled glutamate receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0