This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NDE1 SIRNA Assays ›
Gene Symbol:
Nde1 LOC103690118
Gene Name:
nudE neurodevelopment protein 1
nuclear distribution protein nudE homolog 1-like
Gene Aliases:
Nude
-
Chromosome Location:
Chr.10: 860513 - 904595 on Build Rnor_6.0
Chromosome Location:
Chr.10: 12817 - 24824 on Build Rnor_6.0
UniGene ID:
Rn.20222
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID 54728

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Nde1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053347.1 NP_445799.1 1, 2 91
XM_006245849.2 XP_006245911.1 2, 3 208
XM_006245850.2 XP_006245912.1 3, 4 240
XM_006245851.1 XP_006245913.1 2, 3 208
XM_008767443.1 XP_008765665.1 2, 3 192
XM_008767444.1 XP_008765666.1 2, 3 192
XM_008767445.1 XP_008765667.1 2, 3 192
GenBank mRNA AF240463.1 - 1, 2 91
BC081759.1 - 1, 2 103 7190483
Gene Symbol: LOC103690118
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006245690.2 XP_006245752.1 2, 3 237
XM_008767446.1 XP_008765668.1 3, 4 278

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001285503, NM_001285504, NM_001114085, NM_023317, AK146982, and others.

Gene Ontology Categories:

Function(s) Process(es)

establishment of mitotic spindle orientation
neuron migration
microtubule nucleation
chromosome segregation
mitotic centrosome separation
neuroblast proliferation
cell migration
cerebral cortex development
forebrain development
microtubule organizing center organization
vesicle transport along microtubule
centrosome duplication
cell division
establishment of chromosome localization
centrosome localization
protein binding
microtubule binding
protein domain specific binding
identical protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0