This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other BHLHE40 SIRNA Assays ›
Gene Symbol:
Bhlhe40
Gene Name:
basic helix-loop-helix family, member e40
Gene Aliases:
Bhlhb2, Dec1, SHARP-2, Sharp2, Stra13, Stra14
Chromosome Location:
Chr.4: 140703619 - 140709319 on Build Rnor_6.0
UniGene ID:
Rn.81055
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 54710

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Bhlhe40
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053328.1 NP_445780.1 1, 2 406
GenBank mRNA AF009330.1 - 1, 2 406

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011498, BC010720, AK090032, AF010305, AF364051, and others.

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
transcription from RNA polymerase II promoter
nervous system development
circadian rhythm
response to light stimulus
entrainment of circadian clock
circadian regulation of gene expression
entrainment of circadian clock by photoperiod
negative regulation of sequence-specific DNA binding transcription factor activity
negative regulation of transcription, DNA-templated
regulation of neuronal synaptic plasticity
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II activating transcription factor binding
transcriptional repressor activity, RNA polymerase II transcription factor binding
DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
transcription corepressor activity
protein domain specific binding
protein homodimerization activity
bHLH transcription factor binding
MRF binding
protein heterodimerization activity
protein dimerization activity
E-box binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0