This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GLRX3 SIRNA Assays ›
Gene Symbol:
Glrx3
Gene Name:
glutaredoxin 3
Gene Aliases:
PICOT, Txnl2
Chromosome Location:
Chr.1: 209768696 - 209799912 on Build Rnor_6.0
UniGene ID:
Rn.3578
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 54549

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Glrx3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032614.2 NP_116003.2 4, 5 485
XM_006230497.2 XP_006230559.1 4, 5 898
GenBank mRNA AF118651.1 - 4, 5 319
BC086381.1 - 4, 5 485 7301390
FQ216152.1 - 1, 2 135

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of the force of heart contraction
negative regulation of cardiac muscle hypertrophy
cell redox homeostasis
oxidation-reduction process
protein kinase C binding
electron carrier activity
protein disulfide oxidoreductase activity
poly(A) RNA binding
metal ion binding
iron-sulfur cluster binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0