This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DLL1 SIRNA Assays ›
Gene Symbol:
Dll1
Gene Name:
delta-like 1 (Drosophila)
Gene Aliases:
-
Chromosome Location:
Chr.1: 57318621 - 57326732 on Build Rnor_6.0
UniGene ID:
Rn.10628
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 54527

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dll1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032063.2 NP_114452.1 2 464
GenBank mRNA U78889.2 - 2 464

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_007865, X80903, BC065063, BC057400.

Gene Ontology Categories:

Function(s) Process(es)

cell fate determination
somitogenesis
somite specification
heart looping
Notch signaling pathway
cell-cell signaling
determination of left/right symmetry
compartment pattern specification
nervous system development
auditory receptor cell fate commitment
negative regulation of interleukin-10 production
negative regulation of cell differentiation
negative regulation of auditory receptor cell differentiation
negative regulation of myeloid cell differentiation
negative regulation of neuron differentiation
positive regulation of Notch signaling pathway
positive regulation of transcription from RNA polymerase II promoter
inner ear development
regulation of neurogenesis
left/right axis specification
proximal tubule development
loop of Henle development
neuronal stem cell population maintenance
Notch binding
calcium ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0