This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other UGCG SIRNA Assays ›
Gene Symbol:
Ugcg
Gene Name:
UDP-glucose ceramide glucosyltransferase
Gene Aliases:
-
Chromosome Location:
Chr.5: 76386838 - 76419560 on Build Rnor_6.0
UniGene ID:
Rn.24091
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 54279

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ugcg
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031795.2 NP_113983.1 3 310
GenBank mRNA AF047707.1 - 3 372
AJ224156.1 - 3 296

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

glucosylceramide biosynthetic process
ceramide glucosyltransferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-77574795ff-5hfml:80/100.66.128.142:80.
git-commit: 5cc8a939e1477954742eeb281a87e54f28adfb77
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header