This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other REST SIRNA Assays ›
Gene Symbol:
Rest
Gene Name:
RE1-silencing transcription factor
Gene Aliases:
-
Chromosome Location:
Chr.14: 33131985 - 33157822 on Build Rnor_6.0
UniGene ID:
Rn.10879
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 54183

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rest
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031788.1 NP_113976.1 1 273
XM_006250847.2 XP_006250909.1 352
XM_006250848.1 XP_006250910.1 2 357
XM_006250849.2 XP_006250911.1 2 615
GenBank mRNA AF037199.1 - 1 273
AF037203.1 - 1 273

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
hematopoietic progenitor cell differentiation
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
negative regulation of cell proliferation
negative regulation of gene expression
negative regulation of aldosterone biosynthetic process
cellular response to drug
positive regulation of apoptotic process
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation by host of viral transcription
negative regulation of neuron differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
negative regulation of calcium ion-dependent exocytosis
negative regulation of insulin secretion
negative regulation of neurogenesis
cardiac muscle cell myoblast differentiation
histone H4 deacetylation
cellular response to electrical stimulus
cellular response to glucocorticoid stimulus
potassium ion transmembrane transport
negative regulation of cortisol biosynthetic process
negative regulation of dense core granule biogenesis
negative regulation of mesenchymal stem cell differentiation
negative regulation of amniotic stem cell differentiation
RNA polymerase II transcription factor activity, sequence-specific DNA binding
core promoter sequence-specific DNA binding
core promoter binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
nucleic acid binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription factor binding
outward rectifier potassium channel activity
protein complex binding
transcription regulatory region DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0