This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FOXM1 SIRNA Assays ›
Gene Symbol:
Foxm1
Gene Name:
forkhead box M1
Gene Aliases:
-
Chromosome Location:
Chr.4: 161685283 - 161697633 on Build Rnor_6.0
UniGene ID:
Rn.10665
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 53979

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Foxm1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031633.3 NP_113821.2 2 378
XM_006237416.2 XP_006237478.1 1 351
XM_006237417.2 XP_006237479.1 1 351
XM_006237418.2 XP_006237480.1 2 378
XM_006237419.2 XP_006237481.1 1 351
XM_006237420.2 XP_006237482.1 2 378
XM_006237421.2 XP_006237483.1 2 378
GenBank mRNA CK469502.1 - 1 246
U83112.1 - 2 378

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G2/M transition of mitotic cell cycle
negative regulation of transcription from RNA polymerase II promoter
vasculogenesis
liver development
DNA repair
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
positive regulation of cell proliferation
negative regulation of stress-activated MAPK cascade
response to gonadotropin
regulation of cell proliferation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
regulation of Ras protein signal transduction
response to cAMP
regulation of cell cycle arrest
negative regulation of cell aging
regulation of reactive oxygen species metabolic process
positive regulation of double-strand break repair
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein kinase binding
sequence-specific DNA binding
transcription regulatory region DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header