This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MAPK6 SIRNA Assays ›
Gene Symbol:
Mapk6
Gene Name:
mitogen-activated protein kinase 6
Gene Aliases:
ERK3
Chromosome Location:
Chr.8: 82328013 - 82351108 on Build Rnor_6.0
UniGene ID:
Rn.88457
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 53969

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mapk6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031622.2 NP_113810.2 2 1093
XM_006243409.2 XP_006243471.1 2 1587
GenBank mRNA BC109380.1 - 2 1044 7446401
M64301.1 - 1 721

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_002748, L77964, CR749401, AF420474, AB451301, and others.

Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
protein phosphorylation
MAP kinase activity
ATP binding
protein kinase binding
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0