This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TSC22D3 SIRNA Assays ›
Gene Symbol:
Tsc22d3
Gene Name:
TSC22 domain family, member 3
Gene Aliases:
Dsipi, Gilz
Chromosome Location:
Chr.X: 111884285 - 111944698 on Build Rnor_6.0
UniGene ID:
Rn.21970
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 53778

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tsc22d3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031345.1 NP_112635.1 3 443
XM_006234249.2 XP_006234311.1 4 710
XM_006234250.2 XP_006234312.1 4 624
XM_006234251.2 XP_006234313.1 4 659
XM_008773446.1 XP_008771668.1 3 292
XM_008773447.1 XP_008771669.1 4 563
GenBank mRNA AB025431.1 - 3 443
BC061979.1 - 3 441 5598145

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001077364, NM_010286, AK083389, AF024519, BC028813, and others.

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
response to osmotic stress
body fluid secretion
negative regulation of skeletal muscle tissue development
negative regulation of activation-induced cell death of T cells
transcription factor activity, sequence-specific DNA binding
MRF binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0