This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CAMKK2 SIRNA Assays ›
Gene Symbol:
Camkk2
Gene Name:
calcium/calmodulin-dependent protein kinase kinase 2, beta
Gene Aliases:
-
Chromosome Location:
Chr.12: 39253409 - 39302601 on Build Rnor_6.0
UniGene ID:
Rn.229208
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 53584

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Camkk2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031338.1 NP_112628.1 2, 3 605
GenBank mRNA AB018081.1 - 2, 3 605

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_153499, NM_172216, NM_006549, NM_172214, NM_153500, and others.

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
activation of protein kinase activity
protein autophosphorylation
protein kinase activity
protein serine/threonine kinase activity
calmodulin-dependent protein kinase activity
calcium ion binding
calmodulin binding
ATP binding
protein kinase activator activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header