This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RNPEP SIRNA Assays ›
Gene Symbol:
Rnpep
Gene Name:
arginyl aminopeptidase (aminopeptidase B)
Gene Aliases:
-
Chromosome Location:
Chr.13: 52092866 - 52111237 on Build Rnor_6.0
UniGene ID:
Rn.10979
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 53514

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rnpep
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031097.1 NP_112359.1 3 656
GenBank mRNA AY724503.1 - 3 828
BC061718.1 - 3 686 5598331
D87515.1 - 3 650
U61696.1 - 3 656

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_145417, NM_001159624, AK159445, AK159195, AK145466, and others.

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
aging
protein processing
leukotriene biosynthetic process
negative regulation of blood pressure
retina development in camera-type eye
aminopeptidase activity
copper ion binding
metallopeptidase activity
zinc ion binding
peptide binding
cobalt ion binding
metalloaminopeptidase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0