This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PDPK1 SIRNA Assays ›
Gene Symbol:
Pdpk1
Gene Name:
3-phosphoinositide dependent protein kinase-1
Gene Aliases:
-
Chromosome Location:
Chr.10: 13446373 - 13525248 on Build Rnor_6.0
UniGene ID:
Rn.10905
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 53411

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pdpk1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031081.1 NP_112343.1 4, 5 525
XM_006246036.2 XP_006246098.1 4, 5 2377
XM_006246038.2 XP_006246100.1 4, 5 587
GenBank mRNA Y15748.1 - 4, 5 525

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001286662, NM_011062, NM_001080773, AY062008, BC146001, and others.

Gene Ontology Categories:

Function(s) Process(es)

type B pancreatic cell development
transcription, DNA-templated
regulation of transcription, DNA-templated
protein phosphorylation
negative regulation of protein kinase activity
hyperosmotic response
epidermal growth factor receptor signaling pathway
positive regulation of phospholipase activity
regulation of endothelial cell migration
negative regulation of cardiac muscle cell apoptotic process
cell migration
peptidyl-threonine phosphorylation
calcium-mediated signaling
negative regulation of transforming growth factor beta receptor signaling pathway
activation of protein kinase B activity
cellular response to insulin stimulus
negative regulation of toll-like receptor signaling pathway
intracellular signal transduction
regulation of I-kappaB kinase/NF-kappaB signaling
regulation of mast cell degranulation
focal adhesion assembly
positive regulation of release of sequestered calcium ion into cytosol
cellular response to epidermal growth factor stimulus
positive regulation of establishment of protein localization to plasma membrane
extrinsic apoptotic signaling pathway
protein serine/threonine kinase activity
3-phosphoinositide-dependent protein kinase activity
insulin receptor binding
ATP binding
phospholipase activator activity
kinase activity
protein kinase binding
phospholipase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0