This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ITPKA SIRNA Assays ›
Gene Symbol:
Itpka
Gene Name:
inositol-trisphosphate 3-kinase A
Gene Aliases:
-
Chromosome Location:
Chr.3: 111545007 - 111553605 on Build Rnor_6.0
UniGene ID:
Rn.9877
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 53290

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Itpka
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031045.2 NP_112307.2 5 1127
GenBank mRNA M29787.1 - 5 1127
X56917.1 - 5 1118

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_146125, BC027291.

Gene Ontology Categories:

Function(s) Process(es)

inositol metabolic process
protein phosphorylation
actin cytoskeleton organization
regulation of synaptic plasticity
positive regulation of dendritic spine morphogenesis
dendritic spine maintenance
calmodulin-dependent protein kinase activity
calmodulin binding
ATP binding
inositol-1,4,5-trisphosphate 3-kinase activity
Rac GTPase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0