This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SCN1A SIRNA Assays ›
Gene Symbol:
Scn1a
Gene Name:
sodium channel, voltage-gated, type I, alpha subunit
Gene Aliases:
-
Chromosome Location:
Chr.3: 52388811 - 52533365 on Build Rnor_6.0
UniGene ID:
Rn.32079
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 53149

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Scn1a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030875.1 NP_110502.1 7 1080
XM_008761923.1 XP_008760145.1 7 1100
XM_008761924.1 XP_008760146.1 7 1008
XM_008761925.1 XP_008760147.1 7 916
XM_008761926.1 XP_008760148.1 7 1102
XM_008761927.1 XP_008760149.1 7 1099
XM_008761928.1 XP_008760150.1 7 1100
XM_008761929.1 XP_008760151.1 7 1099
GenBank mRNA M22253.1 - 7 1080
X03638.1 - 7 1080

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

action potential
sodium ion transport
adult walking behavior
neuronal action potential propagation
neuronal action potential
regulation of ion transmembrane transport
sodium ion transmembrane transport
regulation of membrane potential
neuromuscular process controlling posture
membrane depolarization during action potential
voltage-gated sodium channel activity
sodium ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0