This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MVD SIRNA Assays ›
Gene Symbol:
Mvd
Gene Name:
mevalonate (diphospho) decarboxylase
Gene Aliases:
Mvpd
Chromosome Location:
Chr.19: 55258521 - 55269108 on Build Rnor_6.0
UniGene ID:
Rn.10288
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 53124

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mvd
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031062.2 NP_112324.1 2, 3 187
GenBank mRNA BC081784.1 - 2, 3 172 7135885
U53706.1 - 2, 3 188

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cholesterol biosynthetic process
positive regulation of cell proliferation
isoprenoid biosynthetic process
isopentenyl diphosphate biosynthetic process, mevalonate pathway
response to drug
diphosphomevalonate decarboxylase activity
ATP binding
Hsp70 protein binding
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0