This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GTF2F2 SIRNA Assays ›
Gene Symbol:
Gtf2f2
Gene Name:
general transcription factor IIF, polypeptide 2
Gene Aliases:
Rap30
Chromosome Location:
Chr.15: 57942057 - 58068892 on Build Rnor_6.0
UniGene ID:
Rn.11000
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 53107

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gtf2f2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031042.3 NP_112304.2 1, 2 186
GenBank mRNA BC091112.1 - 1, 2 186 7381243
D10665.1 - 1, 2 236
FQ232071.1 - 1, 2 225
FQ233324.1 - 1, 2 225
L01267.1 - 1, 2 163

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_026816, AK144858, BC027173.

Gene Ontology Categories:

Function(s) Process(es)

transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
positive regulation of transcription elongation from RNA polymerase II promoter
positive regulation of RNA polymerase II transcriptional preinitiation complex assembly
positive regulation of transcription from RNA polymerase II promoter
molecular_function
DNA binding
ATP binding
ATP-dependent helicase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0